siRNA Details
| Gene Symbol: WDR92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256476.1 | NP_001243405.1 | 6 | 818 | ||
| NM_138458.3 | NP_612467.1 | 6 | 818 | |||
| NR_046234.1 | 6 | 691 | ||||
| GenBank mRNA | AK056303.1 | - | 6 | 782 | ||
| AK058090.1 | - | 6 | 691 | |||
| AL834142.1 | - | 6 | 654 | |||
| AY541587.1 | - | 6 | 793 | |||
| BC014022.2 | - | 6 | 740 | 4309224 | ||
| BC020271.1 | - | 6 | 772 | |||
| BC036904.1 | - | 6 | 628 | |||
| BC066657.1 | - | 6 | 768 | 6164689 | ||

WDR92
Genomic Map