siRNA Details
| Gene Symbol: CMTM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037288.1 | NP_001032365.1 | 2 | 636 | ||
| NM_001288746.1 | NP_001275675.1 | 2 | 636 | |||
| NM_138460.2 | NP_612469.1 | 2 | 636 | |||
| XM_017020953.1 | XP_016876442.1 | 3 | 622 | |||
| XM_017020954.1 | XP_016876443.1 | 3 | 499 | |||
| GenBank mRNA | AF479262.2 | - | 2 | 637 | ||
| AF527413.1 | - | 2 | 419 | |||
| AK094840.1 | - | 1 | 1368 | |||
| AY820135.1 | - | 2 | 192 | |||
| BC013109.1 | - | 2 | 381 | 4214683 | ||

CMTM5
Genomic Map