siRNA Details
| Gene Symbol: FAM122A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138333.4 | NP_612206.4 | 1 | 991 | ||
| GenBank mRNA | AA418448.1 | - | 1 | 103 | ||
| AK126379.1 | - | 1 | 932 | |||
| BC013062.1 | - | 1 | 942 | 2901027 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM122A| Gene Symbol: FAM122A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138333.4 | NP_612206.4 | 1 | 991 | ||
| GenBank mRNA | AA418448.1 | - | 1 | 103 | ||
| AK126379.1 | - | 1 | 932 | |||
| BC013062.1 | - | 1 | 942 | 2901027 | ||