siRNA Details
| Gene Symbol: ZMYND19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138462.2 | NP_612471.1 | 5 | 713 | ||
| XM_005266052.4 | XP_005266109.1 | 5 | 1142 | |||
| GenBank mRNA | AJ298882.2 | - | 5 | 721 | ||
| AX766496.1 | - | 5 | 721 | |||
| BC012948.1 | - | 5 | 676 | 4519078 | ||

ZMYND19
Genomic Map