siRNA Details
| Gene Symbol: ACSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318890.1 | NP_001305819.1 | 8, 9 | 1316 | ||
| NM_052956.2 | NP_443188.2 | 7, 8 | 1180 | |||
| XM_006721016.2 | XP_006721079.1 | 7, 8 | 1159 | |||
| XM_006721018.2 | XP_006721081.1 | 5, 6 | 538 | |||
| XM_011545729.2 | XP_011544031.1 | 7, 8 | 1159 | |||
| XM_011545731.2 | XP_011544033.1 | 5, 6 | 1355 | |||
| XM_011545732.2 | XP_011544034.1 | 6, 7 | 597 | |||
| XM_017022915.1 | XP_016878404.1 | 6, 7 | 2007 | |||
| GenBank mRNA | AB059429.1 | - | 7, 8 | 1180 | ||
| AK092574.1 | - | 5, 6 | 538 | |||
| AK314282.1 | - | 8, 9 | 1272 | |||
| BC125178.1 | - | 7, 8 | 1180 | 40121956 | ||

ACSM1
Genomic Map