siRNA Details
| Gene Symbol: TM4SF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184723.1 | NP_001171652.1 | 4 | 588 | ||
| NM_138786.3 | NP_620141.1 | 5 | 764 | |||
| GenBank mRNA | AK313400.1 | - | 5 | 538 | ||
| AL833134.1 | - | 5 | 686 | |||
| BC014339.1 | - | 5 | 640 | 3828466 | ||
| CA454331.1 | - | 4 | 550 | |||

TM4SF18
Genomic Map