siRNA Details
Gene Symbol: RAB39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171998.3 | NP_741995.1 | 1 | 328 | ||
GenBank mRNA | AL834460.1 | - | 1 | 191 | ||
AY052478.1 | - | 1 | 226 | |||
BC009714.1 | - | 1 | 328 | 3900092 |
