siRNA Details
| Gene Symbol: WDR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170710.4 | NP_733828.2 | 30 | 4084 | ||
| NM_181265.3 | NP_851782.3 | 28 | 3916 | |||
| XM_005262725.3 | XP_005262782.1 | 28 | 3583 | |||
| XM_011531582.2 | XP_011529884.1 | 29 | 3925 | |||
| XM_017007686.1 | XP_016863175.1 | 29 | 3913 | |||
| XM_017007687.1 | XP_016863176.1 | 30 | 3998 | |||
| XM_017007688.1 | XP_016863177.1 | 30 | 3915 | |||
| XM_017007689.1 | XP_016863178.1 | 28 | 3891 | |||
| XM_017007690.1 | XP_016863179.1 | 29 | 3977 | |||
| XM_017007691.1 | XP_016863180.1 | 28 | 3747 | |||
| XM_017007692.1 | XP_016863181.1 | 29 | 3833 | |||
| XM_017007693.1 | XP_016863182.1 | 27 | 3726 | |||
| XM_017007694.1 | XP_016863183.1 | 28 | 3710 | |||
| XM_017007695.1 | XP_016863184.1 | 28 | 3761 | |||
| XM_017007696.1 | XP_016863185.1 | 27 | 3740 | |||
| GenBank mRNA | AF492460.1 | - | 30 | 3937 | ||
| AK130128.1 | - | 14 | 1546 | |||
| BX648508.1 | - | 14 | 1484 | |||

WDR17
Genomic Map