siRNA Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079536.1 | NP_001073004.1 | 3 | 734 | ||
| NM_053017.4 | NP_443750.2 | 2 | 1047 | |||
| XM_005252779.3 | XP_005252836.1 | 2 | 723 | |||
| XM_006718136.3 | XP_006718199.1 | 3 | 618 | |||
| XM_011519879.2 | XP_011518181.1 | 3 | 768 | |||
| GenBank mRNA | BC014577.1 | - | 2 | 738 | 3934249 | |
| Y16835.1 | - | 3 | 737 | |||

ART5
Genomic Map