siRNA Details
| Gene Symbol: CATSPER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053054.3 | NP_444282.3 | 1 | 1291 | ||
| GenBank mRNA | AF407333.1 | - | 1 | 1153 | ||
| BC032950.2 | - | 1 | 1290 | 4822261 | ||
| BC036522.2 | - | 1 | 1253 | 5268567 | ||

CATSPER1
Genomic Map