siRNA Details
| Gene Symbol: DACH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139514.1 | NP_001132986.1 | 4, 5 | 1067 | ||
| NM_001139515.1 | NP_001132987.1 | 5, 6 | 492 | |||
| NM_053281.3 | NP_444511.1 | 5, 6 | 1106 | |||
| XM_011530846.2 | XP_011529148.1 | 5, 6 | 1105 | |||
| XM_011530847.2 | XP_011529149.1 | 5, 6 | 1105 | |||
| XM_011530848.2 | XP_011529150.1 | 5, 6 | 1105 | |||
| XM_017029254.1 | XP_016884743.1 | 4, 5 | 1066 | |||
| XM_017029255.1 | XP_016884744.1 | 4, 5 | 1066 | |||
| XM_017029256.1 | XP_016884745.1 | 4, 5 | 1066 | |||
| GenBank mRNA | AF428101.1 | - | 5, 6 | 936 | ||
| AK055953.1 | - | 4, 5 | 453 | |||
| AK091842.1 | - | 4, 5 | 1060 | |||
| AK290596.1 | - | 5, 6 | 1106 | |||
| AK298787.1 | - | 5, 6 | 492 | |||
| BC114950.1 | - | 5, 6 | 1015 | 40006598 | ||

DACH2
Genomic Map