siRNA Details
| Gene Symbol: SSX2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166293.1 | NP_001159765.1 | 2 | 289 | ||
| NM_001166417.1 | NP_001159889.1 | 3 | 467 | |||
| NM_014021.3 | NP_054740.3 | 3 | 339 | |||
| XM_005270427.1 | XP_005270484.1 | 2 | 228 | |||
| XM_005270428.1 | XP_005270485.1 | 2 | 186 | |||
| XM_005270429.1 | XP_005270486.1 | 2 | 228 | |||
| XM_011540610.1 | XP_011538912.1 | 3 | 236 | |||
| XM_011540612.2 | XP_011538914.1 | 3 | 277 | |||
| XM_017000234.1 | XP_016855723.1 | 2 | 217 | |||
| XM_017000235.1 | XP_016855724.1 | 2 | 195 | |||
| XM_017000236.1 | XP_016855725.1 | 2 | 211 | |||
| GenBank mRNA | AB023140.1 | - | 2 | 277 | ||
| AK021515.1 | - | 1 | 109 | |||
| AK093173.1 | - | 3 | 236 | |||
| AK292475.1 | - | 2 | 177 | |||
| AL133046.1 | - | 3 | 279 | |||
| AY367055.1 | - | 3 | 202 | |||
| BC033637.1 | - | 2 | 164 | 5578582 | ||
| BC035580.1 | - | 1 | 98 | |||
| BC064389.1 | - | 2 | 253 | 5745142 | ||

SSX2IP
Genomic Map