siRNA Details
| Gene Symbol: SSX2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166293.1 | NP_001159765.1 | 3 | 378 | ||
| NM_001166294.1 | NP_001159766.1 | 2 | 246 | |||
| NM_001166295.1 | NP_001159767.1 | 2 | 374 | |||
| NM_001166417.1 | NP_001159889.1 | 4 | 556 | |||
| NM_014021.3 | NP_054740.3 | 4 | 428 | |||
| XM_005270427.1 | XP_005270484.1 | 3 | 317 | |||
| XM_005270428.1 | XP_005270485.1 | 3 | 275 | |||
| XM_005270429.1 | XP_005270486.1 | 3 | 317 | |||
| XM_006710331.1 | XP_006710394.1 | 3 | 235 | |||
| XM_011540610.1 | XP_011538912.1 | 4 | 325 | |||
| XM_011540612.2 | XP_011538914.1 | 4 | 366 | |||
| XM_017000234.1 | XP_016855723.1 | 3 | 306 | |||
| XM_017000235.1 | XP_016855724.1 | 3 | 284 | |||
| XM_017000236.1 | XP_016855725.1 | 3 | 300 | |||
| XM_017000237.1 | XP_016855726.1 | 2 | 89 | |||
| GenBank mRNA | AB023140.1 | - | 3 | 366 | ||
| AK021515.1 | - | 2 | 198 | |||
| AK093173.1 | - | 4 | 313 | |||
| AK292475.1 | - | 3 | 266 | |||
| AK294057.1 | - | 2 | 133 | |||
| AK316472.1 | - | 2 | 152 | |||
| AL133046.1 | - | 4 | 368 | |||
| AY367055.1 | - | 4 | 291 | |||
| BC033637.1 | - | 3 | 253 | 5578582 | ||
| BC035580.1 | - | 2 | 187 | |||
| BC064389.1 | - | 3 | 342 | 5745142 | ||

SSX2IP
Genomic Map