siRNA Details
| Gene Symbol: GALNT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319052.1 | NP_001305981.1 | 4, 5 | 505 | ||
| NM_054110.4 | NP_473451.3 | 9, 10 | 2246 | |||
| XM_017005682.1 | XP_016861171.1 | 9, 10 | 9659 | |||
| XM_017005683.1 | XP_016861172.1 | 9, 10 | 1809 | |||
| GenBank mRNA | AB078149.1 | - | 9, 10 | 1771 | ||
| AJ505964.1 | - | 3, 4 | 379 | |||
| AK094780.1 | - | 12, 13 | 1819 | |||
| AK312425.1 | - | 9, 10 | 2247 | |||
| AL831925.1 | - | 9, 10 | 2274 | |||
| AL832575.2 | - | 9, 10 | 1748 | |||
| AY035399.1 | - | 9, 10 | 2054 | |||
| AY358443.1 | - | 9, 10 | 2232 | |||
| BC014789.1 | - | 9, 10 | 2196 | 4212702 | ||
| BC030625.1 | - | 4, 5 | 510 | |||
| BQ005863.1 | - | 1, 2 | 358 | |||

GALNT15
Genomic Map