siRNA Details
| Gene Symbol: IP6K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142883.1 | NP_001136355.1 | 3 | 679 | ||
| NM_054111.4 | NP_473452.2 | 2 | 550 | |||
| XM_005248842.3 | XP_005248899.1 | 3 | 459 | |||
| XM_005248843.3 | XP_005248900.1 | 1 | 320 | |||
| XM_011514295.2 | XP_011512597.1 | 2 | 562 | |||
| GenBank mRNA | AF393812.1 | - | 1 | 245 | ||
| AK056586.2 | - | 2 | 470 | |||
| AK223572.1 | - | 2 | 473 | |||
| AK289935.1 | - | 3 | 599 | |||
| BC089389.1 | - | 2 | 550 | 30530548 | ||

IP6K3
Genomic Map