siRNA Details
| Gene Symbol: BORCS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300742.1 | NP_001287671.1 | 3 | 321 | ||
| NM_058169.4 | NP_477517.1 | 3 | 712 | |||
| XM_011520551.2 | XP_011518853.1 | 3 | 355 | |||
| XM_011520552.2 | XP_011518854.1 | 2 | 542 | |||
| XM_011520553.2 | XP_011518855.1 | 3 | 705 | |||
| XM_017018778.1 | XP_016874267.1 | 3 | 705 | |||
| GenBank mRNA | AK075028.1 | - | 3 | 670 | ||
| AY037865.1 | - | 3 | 373 | |||
| AY037866.1 | - | 2 | 229 | |||
| BC013668.1 | - | 3 | 596 | 4154039 | ||
| BP260903.1 | - | 3 | 322 | |||

BORCS5
Genomic Map