siRNA Details
| Gene Symbol: CHCHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203298.2 | NP_976043.1 | 2, 3 | 279 | ||
| GenBank mRNA | AK098720.1 | - | 2 | 916 | ||
| BC015387.1 | - | 2, 3 | 259 | 4400374 | ||
| BC020852.1 | - | 2, 3 | 266 | 4242977 | ||
| BM787401.1 | - | 2, 3 | 280 | |||

CHCHD1
Genomic Map