siRNA Details
| Gene Symbol: MSS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024593.1 | NP_001019764.1 | 3, 4 | 439 | ||
| GenBank mRNA | AK096884.1 | - | 3, 4 | 440 | ||
| AK309870.1 | - | 4, 5 | 495 | |||
| BC094693.1 | - | 3, 4 | 418 | 30529259 | ||
| BC109255.1 | - | 2, 3 | 352 | |||
| BC109256.1 | - | 2, 3 | 352 | |||
| BX537678.1 | - | 3, 4 | 490 | |||
| CR936824.1 | - | 3, 4 | 421 | |||

MSS51
Genomic Map