siRNA Details
| Gene Symbol: PIK3AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152309.2 | NP_689522.2 | 11 | 1828 | ||
| XM_005269498.1 | XP_005269555.1 | 10 | 1351 | |||
| XM_005269499.1 | XP_005269556.1 | 10 | 1375 | |||
| XM_011539248.1 | XP_011537550.1 | 11 | 1820 | |||
| XM_011539249.1 | XP_011537551.1 | 11 | 1820 | |||
| GenBank mRNA | AK092883.1 | - | 10 | 1351 | ||
| AK125635.1 | - | 11 | 1820 | |||
| BC029917.1 | - | 4 | 679 | 5168493 | ||
| BX648550.1 | - | 11 | 1724 | |||

PIK3AP1
Genomic Map