siRNA Details
| Gene Symbol: MORN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098831.1 | NP_001092301.1 | 5 | 486 | ||
| NM_178832.3 | NP_849154.1 | 5 | 575 | |||
| XM_011539250.2 | XP_011537552.1 | 4 | 609 | |||
| XM_011539251.2 | XP_011537553.1 | 5 | 559 | |||
| GenBank mRNA | AJ312050.1 | - | 4 | 478 | ||
| AJ431726.1 | - | 5 | 486 | |||
| AK098791.1 | - | 4 | 378 | |||
| AL834132.1 | - | 5 | 468 | |||
| BC040015.1 | - | 5 | 438 | 5529065 | ||
| BC093023.1 | - | 5 | 479 | 30519946 | ||
| DQ338436.1 | - | 5 | 464 | |||

MORN4
Genomic Map