siRNA Details
| Gene Symbol: COMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144589.2 | NP_653190.2 | 7 | 857 | ||
| GenBank mRNA | AK074421.1 | - | 7 | 810 | ||
| AK290452.1 | - | 7 | 911 | |||
| AK302283.1 | - | 7 | 767 | |||
| AY358476.1 | - | 7 | 856 | |||
| BC023663.2 | - | 7 | 778 | 4897624 | ||
| BC047774.1 | - | 7 | 857 | 5761696 | ||

COMTD1
Genomic Map