siRNA Details
| Gene Symbol: ANKRD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144590.2 | NP_653191.2 | 5 | 788 | ||
| GenBank mRNA | AK313969.1 | - | 5 | 560 | ||
| BC021671.1 | - | 5 | 575 | 3682099 | ||
| BM785517.1 | - | 5 | 631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD22| Gene Symbol: ANKRD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144590.2 | NP_653191.2 | 5 | 788 | ||
| GenBank mRNA | AK313969.1 | - | 5 | 560 | ||
| BC021671.1 | - | 5 | 575 | 3682099 | ||
| BM785517.1 | - | 5 | 631 | |||