siRNA Details
Gene Symbol: ANKRD22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144590.2 | NP_653191.2 | 6 | 920 | ||
GenBank mRNA | AK313969.1 | - | 6 | 692 | ||
BC021671.1 | - | 6 | 707 | 3682099 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ANKRD22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144590.2 | NP_653191.2 | 6 | 920 | ||
GenBank mRNA | AK313969.1 | - | 6 | 692 | ||
BC021671.1 | - | 6 | 707 | 3682099 |