siRNA Details
| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173791.4 | NP_776152.1 | 5 | 3292 | ||
| XM_005269518.3 | XP_005269575.1 | 4 | 3142 | |||
| GenBank mRNA | AL122051.1 | - | 1 | 1129 | ||
| AY211924.1 | - | 1 | 34 | |||
| BC028375.1 | - | 5 | 3254 | 4837939 | ||

PDZD8
Genomic Map