siRNA Details
| Gene Symbol: OR51A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004749.1 | NP_001004749.1 | 1 | 280 | ||
| GenBank mRNA | BC137062.1 | - | 1 | 317 | 9021059 | |
| BC137063.1 | - | 1 | 317 | 9021060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR51A7| Gene Symbol: OR51A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004749.1 | NP_001004749.1 | 1 | 280 | ||
| GenBank mRNA | BC137062.1 | - | 1 | 317 | 9021059 | |
| BC137063.1 | - | 1 | 317 | 9021060 | ||