siRNA Details
| Gene Symbol: OR5P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153445.1 | NP_703146.1 | 1 | 209 | ||
| GenBank mRNA | AF158377.1 | - | 1 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5P3| Gene Symbol: OR5P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153445.1 | NP_703146.1 | 1 | 209 | ||
| GenBank mRNA | AF158377.1 | - | 1 | 209 | ||