siRNA Details
| Gene Symbol: TMEM52B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079815.1 | NP_001073283.1 | 4, 5 | 473 | ||
| NM_153022.3 | NP_694567.1 | 2, 3 | 699 | |||
| XM_006719029.2 | XP_006719092.1 | 3, 4 | 743 | |||
| GenBank mRNA | AK055728.1 | - | 2, 3 | 625 | ||
| AY358845.1 | - | 2, 3 | 656 | |||
| BC098571.1 | - | 3, 4 | 183 | 30921470 | ||
| BC131523.1 | - | 3, 4 | 187 | 40032531 | ||
| DA156941.1 | - | 4, 5 | 530 | |||

TMEM52B
Genomic Map