siRNA Details
| Gene Symbol: TMEM52B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079815.1 | NP_001073283.1 | 5 | 604 | ||
| NM_153022.3 | NP_694567.1 | 3 | 830 | |||
| XM_006719029.2 | XP_006719092.1 | 4 | 874 | |||
| GenBank mRNA | AK055728.1 | - | 3 | 756 | ||
| AY358845.1 | - | 3 | 787 | |||
| BC098571.1 | - | 4 | 314 | 30921470 | ||
| BC131523.1 | - | 4 | 318 | 40032531 | ||

TMEM52B
Genomic Map