siRNA Details
| Gene Symbol: C12orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152318.2 | NP_689531.2 | 4 | 452 | ||
| GenBank mRNA | BC032326.1 | - | 436 | 4385532 | ||
| BG829346.1 | - | 4 | 453 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf45| Gene Symbol: C12orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152318.2 | NP_689531.2 | 4 | 452 | ||
| GenBank mRNA | BC032326.1 | - | 436 | 4385532 | ||
| BG829346.1 | - | 4 | 453 | |||