siRNA Details
| Gene Symbol: OR10AD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004134.1 | NP_001004134.1 | 1 | 398 | ||
| GenBank mRNA | BC136757.1 | - | 1 | 493 | 9020747 | |
| BC136758.1 | - | 1 | 493 | 9020748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10AD1| Gene Symbol: OR10AD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004134.1 | NP_001004134.1 | 1 | 398 | ||
| GenBank mRNA | BC136757.1 | - | 1 | 493 | 9020747 | |
| BC136758.1 | - | 1 | 493 | 9020748 | ||