siRNA Details
| Gene Symbol: GTSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144594.2 | NP_653195.2 | 4 | 233 | ||
| XM_017018800.1 | XP_016874289.1 | 4 | 206 | |||
| GenBank mRNA | AK057504.1 | - | 4 | 1043 | ||
| AK098819.1 | - | 4 | 236 | |||
| BC021179.1 | - | 4 | 199 | 4823487 | ||
| BC105587.1 | - | 4 | 234 | 4697882 | ||

GTSF1
Genomic Map