siRNA Details
| Gene Symbol: FOXN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213596.2 | NP_998761.2 | 10 | 1643 | ||
| XM_011537922.2 | XP_011536224.1 | 10 | 1832 | |||
| XM_017018818.1 | XP_016874307.1 | 8 | 2618 | |||
| GenBank mRNA | AK131519.1 | - | 6 | 1250 | ||
| BC113073.1 | - | 3 | 643 | |||
| BC146825.1 | - | 10 | 1644 | 8860453 | ||

FOXN4
Genomic Map