siRNA Details
| Gene Symbol: FOXN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213596.2 | NP_998761.2 | 9 | 1316 | ||
| XM_011537922.2 | XP_011536224.1 | 9 | 1505 | |||
| XM_017018818.1 | XP_016874307.1 | 7 | 2291 | |||
| GenBank mRNA | AF425596.1 | - | 7 | 1091 | ||
| AF425597.1 | - | 6 | 966 | |||
| AK131519.1 | - | 5 | 923 | |||
| BC113073.1 | - | 2 | 316 | |||
| BC146825.1 | - | 9 | 1317 | 8860453 | ||

FOXN4
Genomic Map