siRNA Details
| Gene Symbol: TEX26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152325.1 | NP_689538.1 | 3 | 303 | ||
| XM_011534919.2 | XP_011533221.1 | 3 | 363 | |||
| XM_011534921.2 | XP_011533223.1 | 3 | 367 | |||
| XM_011534924.2 | XP_011533226.1 | 3 | 367 | |||
| XM_011534925.2 | XP_011533227.1 | 3 | 367 | |||
| XM_011534926.2 | XP_011533228.1 | 3 | 367 | |||
| XM_011534927.2 | XP_011533229.1 | 3 | 367 | |||
| GenBank mRNA | BC030277.1 | - | 3 | 303 | 5167551 | |

TEX26
Genomic Map