siRNA Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173846.4 | NP_776245.2 | 4 | 633 | ||
| XM_005267324.4 | XP_005267381.1 | 4 | 694 | |||
| XM_017020966.1 | XP_016876455.1 | 4 | 597 | |||
| GenBank mRNA | AK098680.1 | - | 4 | 842 | ||
| AW135537.1 | - | 1 | 176 | |||
| AY072034.1 | - | 4 | 570 | |||
| AY173946.1 | - | 4 | 570 | |||
| BC027610.1 | - | 3 | 895 | |||
| BC038970.2 | - | 3 | 522 | 5169309 | ||

TPPP2
Genomic Map