siRNA Details
| Gene Symbol: TARSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152334.2 | NP_689547.2 | 3 | 497 | ||
| XM_006720386.3 | XP_006720449.1 | 3 | 659 | |||
| XM_017021912.1 | XP_016877401.1 | 3 | 659 | |||
| XM_017021913.1 | XP_016877402.1 | 1 | 71 | |||
| GenBank mRNA | AK056653.1 | - | 3 | 487 | ||
| AL833188.1 | - | 3 | 163 | |||
| BC132671.1 | - | 3 | 465 | 40146693 | ||
| BC136315.1 | - | 3 | 465 | 9020302 | ||
| BC143943.1 | - | 3 | 465 | 9052461 | ||

TARSL2
Genomic Map