siRNA Details
| Gene Symbol: DCUN1D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173475.3 | NP_775746.1 | 3 | 743 | ||
| GenBank mRNA | AK123719.1 | - | 3 | 674 | ||
| AY364247.1 | - | 2 | 483 | |||
| BC040442.1 | - | 3 | 723 | 5590225 | ||
| BX648998.1 | - | 3 | 625 | |||

DCUN1D3
Genomic Map