siRNA Details
| Gene Symbol: C16orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144602.3 | NP_653203.1 | 5 | 874 | ||
| GenBank mRNA | AI478870.1 | - | 1 | 227 | ||
| BC021181.2 | - | 5 | 785 | 5268227 | ||
| BC021718.2 | - | 5 | 785 | 5265370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf78| Gene Symbol: C16orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144602.3 | NP_653203.1 | 5 | 874 | ||
| GenBank mRNA | AI478870.1 | - | 1 | 227 | ||
| BC021181.2 | - | 5 | 785 | 5268227 | ||
| BC021718.2 | - | 5 | 785 | 5265370 | ||