siRNA Details
| Gene Symbol: ANKRD26P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026556.1 | 4, 5 | 836 | |||
| GenBank mRNA | BC070117.1 | - | 4, 5 | 836 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD26P1| Gene Symbol: ANKRD26P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026556.1 | 4, 5 | 836 | |||
| GenBank mRNA | BC070117.1 | - | 4, 5 | 836 | ||