siRNA Details
| Gene Symbol: ZG16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145252.2 | NP_660295.2 | 4 | 392 | ||
| GenBank mRNA | AK311810.1 | - | 4 | 260 | ||
| AY359021.1 | - | 4 | 334 | |||
| BC009722.1 | - | 4 | 332 | 3879368 | ||
| BM786200.1 | - | 2 | 101 | |||
| BQ924772.1 | - | 4 | 395 | |||
| EF067313.1 | - | 4 | 291 | |||
| JN411265.1 | - | 4 | 333 | |||

ZG16B
Genomic Map