siRNA Details
| Gene Symbol: ZC3H18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294340.1 | NP_001281269.1 | 2 | 579 | ||
| NM_144604.3 | NP_653205.3 | 2 | 579 | |||
| XM_011522863.1 | XP_011521165.1 | 2 | 579 | |||
| XM_017022934.1 | XP_016878423.1 | 2 | 579 | |||
| GenBank mRNA | AK225807.1 | - | 2 | 549 | ||
| AK300254.1 | - | 2 | 557 | |||
| BC050463.1 | - | 2 | 526 | 6042036 | ||
| CN299704.1 | - | 1 | 213 | |||
| CN299707.1 | - | 2 | 538 | |||

ZC3H18
Genomic Map