siRNA Details
| Gene Symbol: ZPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198844.2 | NP_942141.2 | 5 | 740 | ||
| NM_199321.2 | NP_955353.1 | 6 | 806 | |||
| XM_011524298.1 | XP_011522600.1 | 6 | 844 | |||
| GenBank mRNA | AK292383.1 | - | 6 | 844 | ||
| AY251603.1 | - | 6 | 789 | |||
| AY251604.1 | - | 5 | 723 | |||
| BC043152.2 | - | 5 | 726 | 5297840 | ||

ZPBP2
Genomic Map