siRNA Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099225.1 | NP_001092695.1 | 4 | 600 | ||
| NM_144609.2 | NP_653210.2 | 5 | 659 | |||
| GenBank mRNA | AK056357.1 | - | 5 | 624 | ||
| AK098464.1 | - | 1 | 132 | |||
| AL833590.1 | - | 4 | 562 | |||
| BC023298.1 | - | 5 | 620 | |||
| BC047776.2 | - | 5 | 634 | 6023964 | ||
| BG706097.1 | - | 4 | 611 | |||
| BX648868.1 | - | 4 | 582 | |||

CCDC43
Genomic Map