siRNA Details
| Gene Symbol: CNTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173478.2 | NP_775749.2 | 3 | 571 | ||
| XM_005257043.4 | XP_005257100.1 | 3 | 334 | |||
| XM_005257044.4 | XP_005257101.1 | 3 | 352 | |||
| XM_005257045.4 | XP_005257102.1 | 2 | 494 | |||
| XM_006721690.2 | XP_006721753.1 | 3 | 571 | |||
| XM_011524310.2 | XP_011522612.1 | 3 | 518 | |||
| XM_011524311.2 | XP_011522613.1 | 2 | 176 | |||
| XM_017024171.1 | XP_016879660.1 | 2 | 494 | |||
| GenBank mRNA | AK097456.1 | - | 3 | 487 | ||
| AK302094.1 | - | 3 | 304 | |||
| BC026187.2 | - | 3 | 615 | 5267540 | ||

CNTD1
Genomic Map