siRNA Details
Gene Symbol: TMEM132E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304438.1 | NP_001291367.1 | 4 | 1502 | ||
GenBank mRNA | BC020591.1 | - | 2 | 157 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM132E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304438.1 | NP_001291367.1 | 4 | 1502 | ||
GenBank mRNA | BC020591.1 | - | 2 | 157 |