siRNA Details
| Gene Symbol: C17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142798.2 | NP_001136270.1 | 3, 4 | 350 | ||
| NM_001142799.2 | NP_001136271.1 | 2, 3 | 248 | |||
| NM_174893.3 | NP_777553.1 | 3, 4 | 350 | |||
| GenBank mRNA | AK055800.1 | - | 2 | 1569 | ||
| AK295795.1 | - | 3, 4 | 305 | |||
| BC040036.1 | - | 3, 4 | 282 | 5406966 | ||
| BM011108.1 | - | 3, 4 | 313 | |||
| BU596784.1 | - | 3, 4 | 353 | |||
| BU730728.1 | - | 3 | 563 | |||
| CB119197.1 | - | 3, 4 | 333 | |||
| CN280697.1 | - | 2, 3 | 216 | |||
| Gene Symbol: LOC284112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098506.1 | - | 1 | 3151 | ||
| Gene Symbol: RNASEK-C17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037717.1 | 5, 6 | 1360 | |||
| GenBank mRNA | BC040148.1 | - | 5, 6 | 1117 | 5588971 | |

C17orf49
Genomic Map