siRNA Details
Gene Symbol: SLC5A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042450.2 | NP_001035915.1 | 6 | 536 | ||
NM_001270648.1 | NP_001257577.1 | 6 | 536 | |||
NM_001270649.1 | NP_001257578.1 | 6 | 536 | |||
NM_001282417.1 | NP_001269346.1 | 6 | 567 | |||
NM_152351.4 | NP_689564.3 | 6 | 536 | |||
XM_011523652.1 | XP_011521954.1 | 6 | 692 | |||
XM_011523653.2 | XP_011521955.1 | 6 | 697 | |||
XM_017024191.1 | XP_016879680.1 | 6 | 693 | |||
GenBank mRNA | AK057946.1 | - | 6 | 537 | ||
AK298345.1 | - | 6 | 522 | |||
BC034380.1 | - | 6 | 481 | |||
BC039868.1 | - | 6 | 584 | 4157703 | ||
BC062617.1 | - | 6 | 490 | |||
BP332461.1 | - | 6 | 537 | |||
DR003789.1 | - | 6 | 536 |
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024953.1 | XP_016880442.1 | 6 | 4985 |
