siRNA Details
| Gene Symbol: NLRP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153447.4 | NP_703148.4 | 12 | 3100 | ||
| GenBank mRNA | AY054986.1 | - | 12 | 3100 | ||
| AY154460.1 | - | 12 | 3100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NLRP5| Gene Symbol: NLRP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153447.4 | NP_703148.4 | 12 | 3100 | ||
| GenBank mRNA | AY054986.1 | - | 12 | 3100 | ||
| AY154460.1 | - | 12 | 3100 | |||