siRNA Details
| Gene Symbol: GIPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133261.2 | NP_573568.1 | 4 | 640 | ||
| XM_005259492.3 | XP_005259549.1 | 4 | 680 | |||
| GenBank mRNA | AB073738.1 | - | 4 | 640 | ||
| BC117312.1 | - | 4 | 638 | 40125863 | ||
| BC117314.1 | - | 4 | 638 | 40125865 | ||

GIPC3
Genomic Map