siRNA Details
| Gene Symbol: TCHHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008536.1 | NP_001008536.1 | 3 | 737 | ||
| GenBank mRNA | AY456639.1 | - | 3 | 737 | ||
| BC137493.1 | - | 3 | 727 | 9021493 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCHHL1| Gene Symbol: TCHHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008536.1 | NP_001008536.1 | 3 | 737 | ||
| GenBank mRNA | AY456639.1 | - | 3 | 737 | ||
| BC137493.1 | - | 3 | 727 | 9021493 | ||